Merck
[Rotenoids from Derris trifoliata].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2007-10-09)
Lu-Rong Xu, Shan Li, Jun Wu, Si Zhang
ABSTRACT
Five rotenoids named dehydrodeguelin, dehydrorotenone, 12a-hydroxy rotenone, tephrosin and 12alphabeta-hydroxyrot-2'-enonic acid were isolated from aerial parts of Derris trifoliata. All these compounds were isolated from this plant for the first time.
Sigma-Aldrich
Tephrosin