Merck
  • Dopamine-induced recruitment of dopamine D1 receptors to the plasma membrane.
Dopamine-induced recruitment of dopamine D1 receptors to the plasma membrane.
Proceedings of the National Academy of Sciences of the United States of America (1998-05-20)
H Brismar, M Asghar, R M Carey, P Greengard, A Aperia
ABSTRACT
The recruitment of G protein-coupled receptors from the cytoplasm to the plasma membrane generally is believed to be a constitutive process. We show here by the use of both confocal microscopy and subcellular fractionation that, for at least one such receptor, this recruitment is regulated and not constitutive. Cells from a proximal tubular-like cell line, LLCPK1 cells, were incubated with either a D1 agonist, a dopamine precursor, or an inhibitor of dopamine metabolism to increase dopamine availability in the cell. Each of the three procedures led to a rapid translocation of dopamine D1 receptors from the cytosol to the plasma membrane.
Sigma-Aldrich
Nitecapone, ≥98% (HPLC)