Merck
  • Dislocation of preformed antibiotic-loaded cement spacers (Spacer-G): etiological factors and clinical prognosis.
Dislocation of preformed antibiotic-loaded cement spacers (Spacer-G): etiological factors and clinical prognosis.
The Journal of arthroplasty (2013-11-26)
Guillem Bori, Ester García-Oltra, Alex Soriano, José Rios, Xavier Gallart, Sebastian Garcia
ABSTRACT
This study assesses the factors associated with the dislocation of the Spacer-G and its clinical prognosis. Seventy-four spacers were reviewed. Acetabular bone defects, proximal femoral cementation of the spacer and its relationship to the size of the head spacer were not associated with dislocation. The only variable that it was possible to associate with dislocation was when the previous stem, prior to the spacer placement, was a cemented stem. In patients who experienced a dislocation, infection was not cured during the interim period more frequently than patients who had not experienced a dislocation (P = 0.001) and the final clinical hip evaluation was also worse (P < 0.001). The study concludes that the surgeon should assess different surgical aspects in order to avoid mechanical complications such as dislocation and its consequences.
Product Description
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 100,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 2,000
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Supelco
Poly(methyl methacrylate), analytical standard, 2,480,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 20,000
Sigma-Aldrich
Poly(methyl methacrylate)
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 8,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 4,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 10,000
Supelco
Poly(methyl methacrylate), analytical standard, suitable for gel permeation chromatography (GPC), 50,000
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic