Merck
  • Transcriptional factor ATF6 is involved in odontoblastic differentiation.
Transcriptional factor ATF6 is involved in odontoblastic differentiation.
Journal of dental research (2014-02-27)
J W Kim, H Choi, B C Jeong, S H Oh, S W Hur, B N Lee, S H Kim, J E Nör, J T Koh, Y C Hwang
ABSTRACT
ATF6 is an endoplasmic reticulum (ER) membrane-bound transcription factor that regulates various cellular functions. The purpose of this study was to investigate the role of ATF6 in odontoblast differentiation. Rat tooth germs were isolated, changes in gene expression were evaluated over time, and localization of ATF6 was determined by immunohistochemistry. Human dental pulp cells (HDPCs) were cultured with 50 µg/mL ascorbic acid and 5 mmol/L β-glycerophosphate or 100 ng/mL bone morphogenetic protein 2 to induce differentiation. Translocation of ATF6 was observed by immunofluorescence and confocal microscopy. Overexpression of ATF6 was performed with an adenoviral vector. Matrix mineralization was evaluated by alizarin red staining. Immunoreactivity to anti-ATF6 was observed in the odontoblastic layer of the molar tooth germ, and expressions of ATF6, dentin sialophosphoprotein (DSPP) and dentin matrix protein 1 (DMP1) increased gradually during tooth germ development. When HDPCs were cultured in differentiation media, ATF6, DSPP, and DMP1 expression increased with the expression of unfolded protein response (UPR) markers, BiP and CHOP. Immunofluorescence results showed that ATF6 protein moved from cytoplasm to nucleus when cells were exposed to differentiation media. Notably, overexpression of ATF6 increased DSPP and DMP1 expression, alkaline phosphatase (ALP) activity, and matrix mineralization in HDPC cultures. Inhibition of ATF6 decreased ALP activity and mineralization. These results suggest that ER membrane-bound transcriptional factor ATF6 may be involved in odontoblastic differentiation.
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, BioUltra, ≥5,700 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, Alkaline from porcine kidney, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from calf intestine, buffered aqueous glycerol solution
Sigma-Aldrich
Phosphatase, Alkaline from Escherichia coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline from calf intestinal mucosa, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, Alkaline shrimp, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Sigma-Aldrich
Phosphatase, Alkaline from human placenta, ≥10 units/mg solid (in glycine buffer)
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein