Merck
  • Highly cis-selective and lead-free hydrogenation of 2-hexyne by a supported Pd catalyst with an ionic-liquid layer.
Highly cis-selective and lead-free hydrogenation of 2-hexyne by a supported Pd catalyst with an ionic-liquid layer.
Chemical communications (Cambridge, England) (2014-07-30)
Frederick Schwab, Natascha Weidler, Martin Lucas, Peter Claus
ABSTRACT
A simple Pd/SiO2 catalyst which was modified with the ionic liquid [BMPL][DCA] gave an excellent yield of 88% towards cis-2-hexene in the stereoselective hydrogenation of 2-hexyne. The catalyst outperforms, even at full conversion, the commonly used lead-poisoned, toxic Lindlar catalyst and supported colloidal-based Pd as well.
Sigma-Aldrich
2-Hexyne, 99%
Pricing and availability is not currently available.