Merck
  • Is the rodent maternal separation model a valid and effective model for studies on the early-life impact on ethanol consumption?
Is the rodent maternal separation model a valid and effective model for studies on the early-life impact on ethanol consumption?
Psychopharmacology (2013-08-29)
Ingrid Nylander, Erika Roman
ABSTRACT
Early-life events can cause long-term neurobiological and behavioural changes with a resultant effect upon reward and addiction processes that enhance risk to develop alcohol use disorders. Maternal separation (MS) is used to study the mediating mechanisms of early-life influences in rodents. In MS studies, the pups are exposed to maternal absence during the first postnatal weeks. The outcome of MS experiments exhibits considerable variation and questions have been raised about the validity of MS models. This short review aims to provide information about experimental conditions that are important to consider when assessing the impact of early-life environment on voluntary ethanol consumption. The results from currently used MS protocols are not uniform. However, studies consistently show that longer separations of intact litters predispose for higher ethanol consumption and/or preference in adult male rats as compared to shorter periods of MS. Studies using individual pup MS paradigms, other controls, low ethanol concentrations, adult females or examining adolescent consumption reported no differences or inconsistent results. There is no "a rodent MS model", there are several models and they generate different results. The compiled literature shows that MS is a model of choice for analysis of early-life effects on voluntary ethanol consumption but there are examples of MS paradigms that are not suitable. These studies emphasize the importance to carefully designed MS experiments to supply the optimal conditions to definitely test the research hypothesis and to be particulate in the interpretation of the outcome.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (5 ampules/kit), 100 mg/dL in H2O, ampule of 5 × 5 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, denatured, denatured, reagent grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethanol, absolute, reag. ISO, reag. Ph. Eur., ≥99.8% (GC), liquid (clear, colorless)
Sigma-Aldrich
Ethanol, puriss., meets analytical specification of Ph. Eur., BP, 96.0-97.2%
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®