Merck
Carbofuran toxicity.
Journal of toxicology and environmental health (1994-12-01)
R C Gupta
ABSTRACT
Carbofuran, an anticholinesterase carbamate, is commonly used as an insecticide, nematicide, and acaricide in agricultural practice throughout the world. Due to its widespread use in agriculture, contamination of food, water, and air has become imminent, and consequently adverse health effects are inevitable in humans, animals, wildlife, and fish. Currently, carbofuran's involvement is most frequently encountered in malicious poisoning. The literature on chemical properties, acute toxicity data, poisoning incidences, pharmacokinetics, and mechanism of toxicity of carbofuran is briefly reviewed. Much emphasis is given to the metabolism of carbofuran, and the impact of carbofuran and its two major metabolites (3-hydroxycarbofuran and 3-ketocarbofuran) on overall toxicity. Biochemical (cholinergic and noncholinergic), hematological, and immunological effects induced by carbofuran are discussed in detail. Carbofuran and/or its major metabolites can cross the placental barrier and produce serious effects on the maternal-placental-fetal unit. Carbofuran's toxicity can be potentiated by simultaneous exposure with other cholinesterase inhibitors. Literature on various biomarkers of carbofuran exposure and on induced adverse health effects is also presented. To date, a combination of atropine and memantine remains the most effective antidotal treatment against acute carbofuran toxicity.
Product Description
Supelco
Carbofuran, PESTANAL®, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Carbofuran, 98%
Pricing and availability is not currently available.