- Preparation of tripyrrane analogues from resorcinol and 2-methylresorcinol for applications in the synthesis of new benziporphyrin systems.
Preparation of tripyrrane analogues from resorcinol and 2-methylresorcinol for applications in the synthesis of new benziporphyrin systems.
Chemical communications (Cambridge, England) (2004-01-23)
Kae Miyake, Timothy D Lash
PMID14737537
ABSTRACT
Acid catalyzed condensation of resorcinol or 2-methylresorcinol with 2 equiv. of an acetoxymethylpyrrole gave bis(pyrrolylmethyl)benzene derivatives in moderate yields; these afforded a series of novel aromatic benziporphyrins using the MacDonald "3 + 1" methodology.