Merck
  • Determination of volatile fatty acids in plasma after ethanolic extraction.
Determination of volatile fatty acids in plasma after ethanolic extraction.
The Biochemical journal (1974-07-01)
C Remesy, C Demigne
ABSTRACT
1. A new rapid micro-method for measuring plasma volatile fatty acids is described. The volatile fatty acids are extracted from plasma with ethanol in the presence of a known quantity of internal standard (sodium isobutyrate). After evaporation of the ethanolic solution of the sodium salts, the residue is dissolved in a dilute solution of orthophosphoric acid to permit analysis by g.l.c. 2. A technique of g.l.c. analysis is described which permits the separation of all the volatile fatty acids from the other plasma constituents at temperatures below 100 degrees C in 5 min. 3. Steam-distillation techniques are unsatisfactory when the acetic acid concentrations in the plasma are below 0.2mm. Heating of a number of plasma constituents in acid conditions gives rise to acetic acid. 4. The binding of volatile fatty acids to plasma proteins was studied; this binding is negligible for acetic acid, but increases with the length of the fatty acid carbon chain. 5. The limits of use of the method and the physiological implications are discussed.
Product Description
Supelco
Metal Packed GC Column (fits Agilent, Configuration "C"), phase 20% TCEP, matrix 80/100 Chromosorb P AW support, L × O.D. × I.D. 15.0 ft (4.6 m) × 1/8 in. × 2.1 mm
Supelco
Metal Packed GC Column (fits Agilent, Configuration "A"), phase 12% UCW-98, matrix 80/100 Chromosorb P AW support, L × O.D. × I.D. 2.0 ft (0.61 m) × 1/8 in. × 2.1 mm
Supelco
Metal Packed GC Column (General Configuration), phase 35% DC-200 (350 cstks), matrix 80/100 Chromosorb P AW support, L × O.D. × I.D. 5.0 ft (1.5 m) × 1/8 in. × 2.1 mm