Merck
  • Phase II study evaluating the efficacy, safety, and pharmacodynamic correlative study of dual antiangiogenic inhibition using bevacizumab in combination with sorafenib in patients with advanced malignant melanoma.
Phase II study evaluating the efficacy, safety, and pharmacodynamic correlative study of dual antiangiogenic inhibition using bevacizumab in combination with sorafenib in patients with advanced malignant melanoma.
Cancer chemotherapy and pharmacology (2014-05-13)
Devalingam Mahalingam, Laeeq Malik, Muralidhar Beeram, Jordi Rodon, Kamalesh Sankhala, Alain Mita, Daniel Benjamin, Norma Ketchum, Joel Michalek, Anthony Tolcher, John Wright, John Sarantopoulos
ABSTRACT
Melanomas are vascular tumors with a high incidence of BRAF mutations driving tumor proliferation. Complete inhibition of vascular endothelial growth factor (VEGF) signaling has potential for enhanced antitumor efficacy. Patients with advanced melanoma and adequate organ function were eligible. Sorafenib was given orally at 200 mg BiD for 5 days every week; bevacizumab was administered 5 mg/kg intravenously every 14 days. The primary objective was to determine clinical biological activity. The secondary objectives were safety, tolerability, and time to progression (TTP). Pharmacodynamic analysis included serum VEGF and soluble VEGF receptor-1 and VEGF receptor-2 performed at baseline, C1D15 and C2D1. The study was terminated during the first stage of a Simon two-stage design, after 14 of planned 21 subjects were enrolled. Of the 14 patients who received treatment, no objective tumor responses were observed. Stable disease (SD) ≥16 weeks was observed in 57 % patients, including three patients with SD lasting ≥1 year. Median TTP was 32 weeks. The most frequently reported drug-related adverse events (AEs) were hand-foot syndrome (57.1 %), fatigue (57.1 %), hypertension (64.3 %), and proteinuria (35.7). Grade 3/4 drug-related AEs were hypertension (14.2 %), hand-foot syndrome, proteinuria, and thrombocytopenia (7 % each). Patients with low VEGF (<300 pg/ml) experienced longer TTP than those with high VEGF [median 50 vs. 15 weeks, p = 0.02). A similar pattern was seen for VEGFR1 and VEGFR2, although it did not reach statistical significance. Combined VEGF/VEGFR blockade using bevacizumab with sorafenib shows clinical activity. The linkage between VEGF levels and time to tumor progression needs further exploration.
Product Description
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Nicotinamide (Niacinamide), analytical standard