Merck
  • Highly efficient solid phase synthesis of oligonucleotide analogs containing phosphorodithioate linkages.
Highly efficient solid phase synthesis of oligonucleotide analogs containing phosphorodithioate linkages.
Nucleic acids research (2000-04-11)
D C Capaldi, D L Cole, V T Ravikumar
ABSTRACT
A triester method for the synthesis of deoxynucleoside phosphorodithioate dimers is described. The phosphorodithioate linkage is introduced using a new dithiophosphorylating reagent DPSE-SP(S)Cl(2)where DPSE = 2-diphenylmethylsilylethyl. This group is removed quickly using tetra-butylammonium fluoride leading to the quantitative formation of phosphorodithioate diesters uncontaminated with the corresponding phosphorothioates. The utility of this group is demonstrated by the synthesis of a penta-decathymidylic acid, [T(PS(2))T(PO(2))](7)T, which contains alternating phosphorodithioate/phosphate diester internucleotide linkages.
Sigma-Aldrich
2-(Methyldiphenylsilyl)ethanol, 99%