Merck
  • Construction of a crown ether-like supramolecular library by conjugation of genetically-encoded peptide linkers displayed on bacteriophage T7.
Construction of a crown ether-like supramolecular library by conjugation of genetically-encoded peptide linkers displayed on bacteriophage T7.
Chemical communications (Cambridge, England) (2014-03-13)
Keisuke Fukunaga, Takaaki Hatanaka, Yuji Ito, Michiko Minami, Masumi Taki
ABSTRACT
By using the 10BASEd-T, we have synthesized a crown ether-like macrocyclic library possessing randomized peptide linkers on bacteriophage T7. Among 1.5 × 10(9) diversities of the supramolecule candidates, we have obtained a specific binder for the N-terminal domain of Hsp90.
Sigma-Aldrich
2,2′-(Ethylenedioxy)bis(ethylamine), 98%
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
SIGMAFAST OPD, tablet