- Unexpected remarkable stability of primary ozonides derived from alkenyl stannanes. One-pot synthesis of 1,2-diols from alkynes.
Unexpected remarkable stability of primary ozonides derived from alkenyl stannanes. One-pot synthesis of 1,2-diols from alkynes.
Organic letters (2002-02-01)
Ana M Gómez, María D Company, Serafín Valverde, J Cristóbal López
PMID11820885
ABSTRACT
Primary ozonides derived from alkenyl stannanes display an unusual stability and can be transformed into 1,2-diols by treatment with dimethyl sulfide and borane-methyl sulfide complex. This observation has been incorporated into the development of a novel one-pot strategy for the conversion of alkynes into 1,2-diols.