Merck
  • Combination therapy with sorafenib and radiofrequency ablation for BCLC Stage 0-B1 hepatocellular carcinoma: a multicenter retrospective cohort study.
Combination therapy with sorafenib and radiofrequency ablation for BCLC Stage 0-B1 hepatocellular carcinoma: a multicenter retrospective cohort study.
The American journal of gastroenterology (2014-11-19)
Xiaobin Feng, Ruocai Xu, Xilin Du, Kefeng Dou, Xiao Qin, Jun Xu, Weidong Jia, Zhiming Wang, Hongzhi Zhao, Shufa Yang, Chenyang Guo, Tianqi Liu, Kuansheng Ma
ABSTRACT
The objective of this study was to evaluate the efficacy of combined therapy using Sorafenib and radiofrequency ablation (RFA) with curative intent for all detectable lesions in patients with Barcelona Clinic Liver Cancer (BCLC) Stage 0-B1 hepatocellular carcinoma (HCC). One hundred and twenty-eight patients with HCC from 12 centers were enrolled in this retrospective study; 64 patients who received Sorafenib plus RFA (Sorafenib-RFA) were compared with a control group treated with RFA alone. The two patient groups were selected with a predefined criterion and matched in terms of their clinical and tumor characteristics at baseline. The primary end point of the study was the incidence of post-RFA HCC recurrence. Secondary end points were overall survival (OS) and treatment toxicity. During a median follow-up of 134.1 weeks, 49 patients died and 79 survived. The 1-, 2-, and 3-year cumulative incidences of post-RFA recurrence were 40.5%, 62.9%, and 74.5%, respectively, in the Sorafenib-RFA group, and 62.8%, 85.4%, and 92.7%, respectively, in the RFA group. The 1-, 2-, 3-, and 4-year OS rates were 85.6%, 64.0%, 58.7%, and 50.3%, respectively, in the Sorafenib-RFA group, and 80.7%, 47.2%, 30.9%, and 30.9%, respectively, in the RFA group. Thus, the Sorafenib-RFA group exhibited better survival than the RFA alone group. Combined therapy with Sorafenib-RFA was associated with a lower incidence of post-RFA recurrence and better OS than RFA alone in patients with BCLC Stage 0-B1 HCC. Although these findings suggest that Sorafenib and RFA is safe and effective for the treatment of early HCC, prospective and randomized controlled trials are needed to validate them.
Product Description
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Niacinamide, meets USP testing specifications
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Nicotinamide (Niacinamide), analytical standard