- Synthesis and antiviral properties of (Z)-5-(2-bromovinyl)-2'-deoxyuridine.
Synthesis and antiviral properties of (Z)-5-(2-bromovinyl)-2'-deoxyuridine.
Journal of medicinal chemistry (1981-06-01)
A S Jones, S G Rahim, R T Walker, E De Clercq
PMID6265638
ABSTRACT
(Z)-5-(2-Bromovinyl)uracil was obtained by photoisomerization of the E. isomer. Similarly, (E)-5-(2-bromovinyl)-2'-deoxyuridine gave the required Z isomer. (Z)-5-(2-Bromovinyl)-2'-deoxyuridine is much less active against herpes simplex virus type 1 (HSV-1) and somewhat less active against herpes simplex virus type 2 than is the E isomer. Both isomers show similar activity against vaccinia virus. Therefore, the highly potent and selective activity of (E)-5-(2-bromovinyl)-2'-deoxyuridine against HSV-1 is due to its E configuration.
MATERIALS