- Micro method for determination of reactive carbonyl groups in proteins and peptides, using 2,4-dinitrophenylhydrazine.
Micro method for determination of reactive carbonyl groups in proteins and peptides, using 2,4-dinitrophenylhydrazine.
The Biochemical journal (1971-02-01)
R Fields, H B Dixon
PMID5114969
ABSTRACT
A method is described for determining carbonyl groups that is especially suitable for use with proteins and peptides. It involves the determination of the extinction at 370nm of a sample solution after adding 2,4-dinitrophenylhydrazine. The reaction of 2,4-dinitrophenylhydrazine with pyruvoylglycine and with transaminated ribonuclease T(1) is presented; the isolation of protein hydrazones is discussed.
Product Description
Supelco
2,4-Dinitrophenylhydrazine hydrochloric acid solution, for TLC derivatization, ~0.005 M in ethanol