Merck
  • Mps1 phosphorylates Borealin to control Aurora B activity and chromosome alignment.
Mps1 phosphorylates Borealin to control Aurora B activity and chromosome alignment.
Cell (2008-02-05)
Nannette Jelluma, Arjan B Brenkman, Niels J F van den Broek, Carin W A Cruijsen, Maria H J van Osch, Susanne M A Lens, René H Medema, Geert J P L Kops
ABSTRACT
Maintenance of chromosomal stability relies on coordination between various processes that are critical for proper chromosome segregation in mitosis. Here we show that monopolar spindle 1 (Mps1) kinase, which is essential for the mitotic checkpoint, also controls correction of improper chromosome attachments. We report that Borealin/DasraB, a member of the complex that regulates the Aurora B kinase, is directly phosphorylated by Mps1 on residues that are crucial for Aurora B activity and chromosome alignment. As a result, cells lacking Mps1 kinase activity fail to efficiently align chromosomes due to impaired Aurora B function at centromeres, leaving improper attachments uncorrected. Strikingly, Borealin/DasraB bearing phosphomimetic mutations restores Aurora B activity and alignment in Mps1-depleted cells. Mps1 thus coordinates attachment error correction and checkpoint signaling, two crucial responses to unproductive chromosome attachments.