Merck
  • Inhibition of bromodomain proteins for the treatment of human diffuse large B-cell lymphoma.
Inhibition of bromodomain proteins for the treatment of human diffuse large B-cell lymphoma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-07-11)
Sally E Trabucco, Rachel M Gerstein, Andrew M Evens, James E Bradner, Leonard D Shultz, Dale L Greiner, Hong Zhang
ABSTRACT
Approximately 50% of patients with diffuse large B-cell lymphoma (DLBCL) enter long-term remission after standard chemotherapy. Patients with DLBCL who do not respond to chemotherapy have few treatment options. There remains a critical need to identify effective and targeted therapeutics for DLBCL. Recent studies have highlighted the incidence of increased c-MYC protein in DLBCL and the correlation between high levels of c-MYC protein and poor survival prognosis of patients with DLBCL, suggesting that c-MYC is a compelling target for DLBCL therapy. The small molecule JQ1 suppresses c-MYC expression through inhibition of the bromodomain and extra-terminal (BET) family of bromodomain proteins. We investigated whether JQ1 can inhibit proliferation of DLBCL cells in culture and xenograft models in vivo. We show that JQ1 at nanomolar concentrations efficiently inhibited proliferation of human DLBCL cells in a dose-dependent manner regardless of their molecular subtypes, suggesting a broad effect of JQ1 in DLBCL. The initial G1 arrest induced by JQ1 treatment in DLBCL cells was followed by either apoptosis or senescence. The expression of c-MYC was suppressed as a result of JQ1 treatment from the natural, chromosomally translocated, or amplified loci. Furthermore, JQ1 treatment significantly suppressed growth of DLBCL cells engrafted in mice and improved survival of engrafted mice. Our results demonstrate that inhibition of the BET family of bromodomain proteins by JQ1 has potential clinical use in the treatment of DLBCL.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide solution, 50 wt. % in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Deoxycholic acid, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Deoxycholic acid, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water