Merck
  • Fennel and raspberry leaf as possible inhibitors of acetaminophen oxidation.
Fennel and raspberry leaf as possible inhibitors of acetaminophen oxidation.
Phytotherapy research : PTR (2014-01-25)
Astrid Jordet Langhammer, Odd Georg Nilsen
ABSTRACT
In addition to CYP2E1, several CYP isoenzymes, notably CYP1A2, 2D6, and 3A4, are suggested to contribute in acetaminophen oxidation and formation of the hepatotoxic metabolite N-acetyl-p-benzoquinone imine (NAPQI). The in vitro CYP2E1 inhibitory potentials of fennel and raspberry leaf, herbs previously found to inhibit CYP1A2, 2D6, and 3A4 activities in vitro, were investigated. Extracts from commercially available herbal products were incubated with recombinant cDNA-expressed human CYP2E1. A validated LC/MS/MS methodology was applied for determination of 6-hydroxychlorzoxazone formation with disulfiram used as a positive inhibitory control. CYP2E1 IC50 inhibition constants were found to be 23 ± 4 and 27 ± 5 µg/ml for fennel and raspberry leaf, respectively, constants significantly lower than those presented in the literature for other herbal extracts. Together with previous findings, the presented in vitro data for CYP2E1 inhibition suggest that fennel and raspberry leaf have a significant potential of inhibiting all the major metabolic pathways for acetaminophen oxidation and NAPQI formation. Both herbs should be further investigated for their in vivo ability of inhibiting acetaminophen oxidation and NAPQI formation.
Product Description
Supelco
Acetaminophen, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Monobasic Potassium Phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetraethylthiuram disulfide, ≥97.0% (S)
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetaminophen, analytical standard
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Chlorzoxazone
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Potassium phosphate dibasic, ACS reagent, ≥98%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate dibasic, reagent grade, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Potassium phosphate dibasic, anhydrous, suitable for luminescence, Molecular Biology, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium phosphate monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Potassium phosphate dibasic solution, 1.0 M
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate dibasic, meets USP testing specifications
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Disulfiram, European Pharmacopoeia (EP) Reference Standard