Merck
  • Harnessing the synthetic capabilities of glycopeptide antibiotic tailoring enzymes: characterization of the UK-68,597 biosynthetic cluster.
Harnessing the synthetic capabilities of glycopeptide antibiotic tailoring enzymes: characterization of the UK-68,597 biosynthetic cluster.
Chembiochem : a European journal of chemical biology (2014-09-27)
Grace Yim, Lindsay Kalan, Kalinka Koteva, Maulik N Thaker, Nicholas Waglechner, Irene Tang, Gerard D Wright
ABSTRACT
In this study, a draft genome sequence of Actinoplanes sp. ATCC 53533 was assembled, and an 81-kb biosynthetic cluster for the unusual sulfated glycopeptide UK-68,597 was identified. Glycopeptide antibiotics are important in the treatment of infections caused by Gram-positive bacteria. Glycopeptides contain heptapeptide backbones that are modified by many tailoring enzymes, including glycosyltransferases, sulfotransferases, methyltransferases, and halogenases, generating extensive chemical and functional diversity. Several tailoring enzymes in the cluster were examined in vitro for their ability to modify glycopeptides, resulting in the synthesis of novel molecules. Tailoring enzymes were also expressed in the producer of the glycopeptide aglycone A47934, generating additional chemical diversity. This work characterizes the biosynthetic program of UK-68,597 and demonstrates the capacity to expand glycopeptide chemical diversity by harnessing the unique chemistry of tailoring enzymes.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Chloramphenicol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Chloramphenicol, γ-irradiated
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Supelco
Chloramphenicol, VETRANAL®, analytical standard
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O
Sigma-Aldrich
Formic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥98%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, puriss., meets analytical specifications of DAC, FCC, 98.0-100%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%