Merck
  • Chitosan microencapsulation of various essential oils to enhance the functional properties of cotton fabric.
Chitosan microencapsulation of various essential oils to enhance the functional properties of cotton fabric.
Journal of microencapsulation (2014-02-06)
Amjed Javid, Zulfiqar Ali Raza, Tanveer Hussain, Asma Rehman
ABSTRACT
The present study dealt with emulsive fabrication of chitosan microcapsules encapsulating essential oils in the present of bio/surfactant. The size distribution, morphology and stability of microcapsules were examined by using advanced surface characterisation techniques. At cetyl trimethyl ammonium bromide (CTAB) concentration of 330 mg/L, the smallest average size of microcapsules was observed as12.8 μm; whereas with biosurfactant at 50 mg/L, the microcapsules of smallest average size of 7.5 μm were observed. The fabricated microcapsules were applied on a desized, bleached and mercerised cotton fabric by using pad-dry-cure method by using a modified dihydroxy ethylene urea as a cross-linking agent. The cross-linking was confirmed by using scanning electron microscopy and Fourier transform infrared spectroscopy techniques. The antibacterial activity of finished fabric was evaluated using the turbidity estimation method. The stiffness and wrinkle recovery properties of the treated fabric were also investigated by using the standard methods. In general, antibacterial activity of treated fabric increased with the increase in chitosan and essential oil concentrations, whereas stiffness increased with increase in concentration of chitosan but decreased with increase in essential oil concentration.
Product Description
USP
Glacial acetic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, Molecular Biology, ≥99.0% (AT)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, Molecular Biology, ≥99%
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, puriss. p.a., ACS reagent, K ≤0.02%, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, purum, ≥32%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%