Merck
  • An LC-MS/MS method for the quantitation of cabozantinib in rat plasma: application to a pharmacokinetic study.
An LC-MS/MS method for the quantitation of cabozantinib in rat plasma: application to a pharmacokinetic study.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-02-14)
Qinghong Su, Jian Li, Xiwei Ji, Jingyun Li, Tianyan Zhou, Wei Lu, Liang Li
ABSTRACT
A simple, rapid and sensitive high-performance liquid chromatography tandem mass-spectrometric method (LC-MS/MS) for the novel multiple tyrosine kinase inhibitor (TKI) cabozantinib was developed and validated using erlotinib as internal standard (IS). Plasma samples were pre-treated by liquid-liquid extraction with ethyl acetate. Separation was achieved on a reversed phase C18 column (50×2mm, 5μm) at ambient temperature using isocratic elution with acetonitrile-water (45:55, v/v) containing 5mM ammonium formate buffer (finally adjusted to apparent pH*=5.0 with formic acid) at a flow rate of 0.4mL/min. The analytes were monitored by a triple quadrupole tandem mass spectrometer with electrospray ionization source in the positive ion mode. Calibration curve was linear (r>0.99) in a concentration range of 0.5-1000ng/mL with the lower limit of quantification (LLOQ) of 0.5ng/mL. Intra- and inter-day accuracy and precision were validated by relative error values (RE) and relative standard deviation values (RSD), respectively, which were both lower than the limit of 15%. This method was successfully applied to a pharmacokinetic study of cabozantinib in rats.
Product Description
Supelco
Ethyl acetate, analytical standard
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Ethyl acetate, ≥99%, FCC, FG
Sigma-Aldrich
Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ammonium formate solution, BioUltra, 10 M in H2O
Supelco
Ammonium formate solution, 10 mM in H2O, suitable for HPLC
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethyl Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl acetate, anhydrous, 99.8%
Sigma-Aldrich
Ethyl acetate, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethyl acetate, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Ethyl acetate, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethyl acetate, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Ethyl acetate, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl acetate, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)