Merck
  • Efficient hole-blocking layer-free planar halide perovskite thin-film solar cells.
Efficient hole-blocking layer-free planar halide perovskite thin-film solar cells.
Nature communications (2015-03-24)
Weijun Ke, Guojia Fang, Jiawei Wan, Hong Tao, Qin Liu, Liangbin Xiong, Pingli Qin, Jing Wang, Hongwei Lei, Guang Yang, Minchao Qin, Xingzhong Zhao, Yanfa Yan
ABSTRACT
Efficient lead halide perovskite solar cells use hole-blocking layers to help collection of photogenerated electrons and to achieve high open-circuit voltages. Here, we report the realization of efficient perovskite solar cells grown directly on fluorine-doped tin oxide-coated substrates without using any hole-blocking layers. With ultraviolet-ozone treatment of the substrates, a planar Au/hole-transporting material/CH₃NH₃PbI₃-xClx/substrate cell processed by a solution method has achieved a power conversion efficiency of over 14% and an open-circuit voltage of 1.06 V measured under reverse voltage scan. The open-circuit voltage is as high as that of our best reference cell with a TiO₂ hole-blocking layer. Besides ultraviolet-ozone treatment, we find that involving Cl in the synthesis is another key for realizing high open-circuit voltage perovskite solar cells without hole-blocking layers. Our results suggest that TiO₂ may not be the ultimate interfacial material for achieving high-performance perovskite solar cells.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methylamine solution, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamine solution, 2.0 M in THF
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
N,N-Dimethylformamide, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Chlorobenzene, ACS reagent, ≥99.5%
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Chlorobenzene, anhydrous, 99.8%
Sigma-Aldrich
Chlorobenzene, ReagentPlus®, 99%
Supelco
Chlorobenzene, analytical standard
Sigma-Aldrich
Chlorobenzene, suitable for HPLC, 99.9%
Sigma-Aldrich
N,N-Dimethylformamide, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamide, ACS reagent, ≥99.8%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Diethyl ether, contains BHT as inhibitor, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free