Merck
  • IgM and IgA rheumatoid factors purified from rheumatoid arthritis sera boost the Fc receptor- and complement-dependent effector functions of the disease-specific anti-citrullinated protein autoantibodies.
IgM and IgA rheumatoid factors purified from rheumatoid arthritis sera boost the Fc receptor- and complement-dependent effector functions of the disease-specific anti-citrullinated protein autoantibodies.
Journal of immunology (Baltimore, Md. : 1950) (2015-03-15)
Florence Anquetil, Cyril Clavel, Géraldine Offer, Guy Serre, Mireille Sebbag
ABSTRACT
Rheumatoid factors (RF) and the disease-specific anti-citrullinated protein autoantibodies (ACPA) coexist in the joints of rheumatoid arthritis (RA) patients where they probably contribute to synovitis. We investigated the influence of IgM and IgA RF on the FcR- and complement-dependent effects of ACPA immune complexes (ACPA-IC). When stimulated by ACPA-IC formed in the presence of IgM RF or IgA RF fractions purified from RA serum pools, M-CSF-generated macrophages skewed their cytokine response toward inflammation, with increases in the TNF-α/IL-10 ratio and in IL-6 and IL-8 secretion, and decreases in the IL-1Ra/IL-1β ratio. In the IgM RF-mediated amplification of the inflammatory response of macrophages, the participation of an IgM receptor was excluded, notably by showing that they did not express any established receptor for IgM. Rather, this amplification depended on the IgM RF-mediated recruitment of more IgG into the ACPA-IC. However, the macrophages expressed FcαRI and blocking its interaction with IgA inhibited the IgA RF-mediated amplification of TNF-α secretion induced by ACPA-IC, showing its major implication in the effects of RF of the IgA class. LPS further amplified the TNF-α response of macrophages to RF-containing ACPA-IC. Lastly, the presence of IgM or IgA RF increased the capacity of ACPA-IC to activate the complement cascade. Therefore, specifically using autoantibodies from RA patients, the strong FcR-mediated or complement-dependent pathogenic potential of IC including both ACPA and IgM or IgA RF was established. Simultaneous FcR triggering by these RF-containing ACPA-IC and TLR4 ligation possibly makes a major contribution to RA synovitis.
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
DL-Cysteine, technical grade
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Supelco
Calcium standard for AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)