Merck
  • Multiple treatments with L-3,4-dihydroxyphenylalanine modulate dopamine biosynthesis and neurotoxicity through the protein kinase A-transient extracellular signal-regulated kinase and exchange protein activation by cyclic AMP-sustained extracellular signal-regulated kinase signaling pathways.
Multiple treatments with L-3,4-dihydroxyphenylalanine modulate dopamine biosynthesis and neurotoxicity through the protein kinase A-transient extracellular signal-regulated kinase and exchange protein activation by cyclic AMP-sustained extracellular signal-regulated kinase signaling pathways.
Journal of neuroscience research (2014-07-22)
Keun Hong Park, Hyun Jin Park, Keon Sung Shin, Myung Koo Lee
ABSTRACT
Multiple treatments with L-3,4-dihydroxyphenylalanine (L-DOPA; 20 µM) induce neurite-like outgrowth and reduce dopamine biosynthesis in rat adrenal pheochromocytoma (PC) 12 cells. We therefore investigated the effects of multiple treatments with L-DOPA (MT-LD) on cell survival and death over a duration of 6 days by using PC12 cells and embryonic rat midbrain primary cell cultures. MT-LD (10 and 20 µM) decreased cell viability, and both types of cells advanced to the differentiation process at 4-6 days. MT-LD induced cyclic adenosine monophosphate (cAMP)-dependent protein kinase A (PKA) phosphorylation and exchange protein activation by cAMP (Epac) expression at 1-3 days, which led to transient extracellular signal-regulated kinase (ERK1/2) phosphorylation in both cells. In these states, MT-LD activated cAMP-response element binding protein (CREB; Ser133) and tyrosine hydroxylase (Ser40) phosphorylation in PC12 cells, which led to an increase in intracellular dopamine levels. In contrast, MT-LD induced prolonged Epac expression at 4-5 days in both cells, which led to sustained ERK1/2 phosphorylation. In these states, the dopamine levels were decreased in PC12 cells. In addition, MT-LD induced c-Jun N-terminal kinase1/2 phosphorylation and cleaved caspase-3 expression at 4-6 days in both cells. These results suggest that MT-LD maintains cell survival via PKA-transient ERK1/2 activation, which stimulates dopamine biosynthesis. In contrast, at the later time period, MT-LD induces differentiation via both prolonged Epac and sustained ERK1/2 activation, which subsequently leads to the cell death process. Our data demonstrate that L-DOPA can cause neurotoxicity by modulating the Epac-ERK pathways in neuronal and PC12 cells.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
3,4-Dihydroxy-L-phenylalanine, ≥98% (TLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
USP
Levodopa, United States Pharmacopeia (USP) Reference Standard
Levodopa, European Pharmacopoeia (EP) Reference Standard
Supelco
Levodopa, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Brefeldin A, Ready Made Solution, from Penicillium brefeldianum, 10 mg/mL in DMSO
Sigma-Aldrich
Brefeldin A, from Penicillium brefeldianum, ≥99% (HPLC and TLC)
Sigma-Aldrich
Brefeldin A, ≥99% (HPLC and TLC), BioXtra, Molecular Biology