Merck
  • Estradiol affects extracellular leptin:adiponectin ratio in human breast tissue in vivo.
Estradiol affects extracellular leptin:adiponectin ratio in human breast tissue in vivo.
The Journal of clinical endocrinology and metabolism (2014-05-07)
Vivian Morad, Annelie Abrahamsson, Charlotta Dabrosin
ABSTRACT
Exposure to sex steroids is associated with increased breast cancer risk, and adipokines, leptin and adiponectin have been implicated in cancer progression. However, it is not known whether sex steroids affect adipokine secretion in breast tissue. To elucidate the role of estrogen and tamoxifen on adipokine release in normal human breast tissue and breast cancer. Microdialysis sampling was used to collect extracellular in vivo leptin and adiponectin from normal human breast tissue in premenopausal healthy volunteers during the menstrual cycle and in postmenopausal women before tamoxifen treatment and after 6 weeks of treatment. In women with breast cancer, microdialysis was performed intratumorally before surgery. In addition, whole normal breast tissue biopsies were cultured ex vivo, and murine breast cancer models were evaluated. In normal breast tissue, plasma estradiol negatively correlated with local extracellular adiponectin levels (r = -0.34; P < .05) and positively correlated with leptin (r = 0.37; P < .05) and leptin:adiponectin ratio (r = 0.38; P < .05). In postmenopausal women, tamoxifen treatment increased adiponectin (P < 0.05) and decreased leptin (P < .01) and the leptin:adiponectin ratio (P < .01). These in vivo results were confirmed in breast tissue biopsies cultured ex vivo. In patients with breast cancer, extracellular leptin was higher (P < .01) and adiponectin lower (P < .05) in tumors than in normal adjacent breast tissue. In a murine model of breast cancer, estrogen exposure increased leptin secretion (P < .05). Estrogen exposure may have a critical role in the regulation of adipokines in human breast tissue and may serve as therapeutic targets for treatment and prevention.
Sigma-Aldrich
Phenol Red, ACS reagent
Supelco
Estradiol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tamoxifen, ≥99%
Supelco
Tamoxifen, analytical standard
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
USP
Estradiol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Estradiol, meets USP testing specifications
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Progesterone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Progesterone, United States Pharmacopeia (USP) Reference Standard
Progesterone, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
Progesterone, VETRANAL®, analytical standard
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Progesterone, ≥99%