Merck
  • Cholecystokinin inhibits inducible nitric oxide synthase expression by lipopolysaccharide-stimulated peritoneal macrophages.
Cholecystokinin inhibits inducible nitric oxide synthase expression by lipopolysaccharide-stimulated peritoneal macrophages.
Mediators of inflammation (2014-08-16)
Rafael Simone Saia, Fabíola Leslie Mestriner, Giuliana Bertozi, Fernando Queiróz Cunha, Evelin Capellari Cárnio
ABSTRACT
Cholecystokinin (CCK) was first described as a gastrointestinal hormone. However, apart from its gastrointestinal effects, studies have described that CCK also plays immunoregulatory roles. Taking in account the involvement of inducible nitric oxide synthase- (iNOS-) derived NO in the sepsis context, the present study was undertaken to investigate the role of CCK on iNOS expression in LPS-activated peritoneal macrophages. Our results revealed that CCK reduces NO production and attenuates the iNOS mRNA expression and protein formation. Furthermore, CCK inhibited the nuclear factor- (NF-) κB pathway reducing IκBα degradation and minor p65-dependent translocation to the nucleus. Moreover, CCK restored the intracellular cAMP content activating the protein kinase A (PKA) pathway, which resulted in a negative modulatory role on iNOS expression. In peritoneal macrophages, the CCK-1R expression, but not CCK-2R, was predominant and upregulated by LPS. The pharmacological studies confirmed that CCK-1R subtype is the major receptor responsible for the biological effects of CCK. These data suggest an anti-inflammatory role for the peptide CCK in modulating iNOS-derived NO synthesis, possibly controlling the macrophage activation through NF-κB, cAMP-PKA, and CCK-1R pathways. Based on these findings, CCK could be used as an adjuvant agent to modulate the inflammatory response and prevent systemic complications commonly found during sepsis.
Product Description
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Supelco
Forskolin, analytical standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard