Merck
  • Chronic heart damage following doxorubicin treatment is alleviated by lovastatin.
Chronic heart damage following doxorubicin treatment is alleviated by lovastatin.
Pharmacological research (2014-12-03)
Christian Henninger, Stefanie Huelsenbeck, Philip Wenzel, Moritz Brand, Johannes Huelsenbeck, Arno Schad, Gerhard Fritz
ABSTRACT
The anticancer efficacy of anthracyclines is limited by cumulative dose-dependent early and delayed cardiotoxicity resulting in congestive heart failure. Mechanisms responsible for anthracycline-induced heart damage are controversially discussed and effective preventive measures are preferable. Here, we analyzed the influence of the lipid lowering drug lovastatin on anthracycline-induced late cardiotoxicity three month after treatment of C57BL/6 mice with five low doses of doxorubicin (5×3mg/kg BW; i.p.). Doxorubicin increased the cardiac mRNA levels of BNP, IL-6 and CTGF, while the expression of ANP remained unchanged. Lovastatin counteracted these persisting cardiac stress responses evoked by the anthracycline. Doxorubicin-induced fibrotic alterations were neither detected by histochemical collagen staining of heart sections nor by analysis of the mRNA expression of collagens. Extensive qRT-PCR-array based analyses revealed a large increase in the mRNA level of heat shock protein Hspa1b in doxorubicin-treated mice, which was mitigated by lovastatin co-treatment. Electron microscopy together with qPCR-based analysis of mitochondrial DNA content indicate that lovastatin attenuates doxorubicin-stimulated hyperproliferation of mitochondria. This was not paralleled by increased expression of oxidative stress responsive genes or senescence-associated proteins. Echocardiographic analyses disclosed that lovastatin protects from the doxorubicin-induced decrease in the left ventricular posterior wall diameter (LVPWD), while constrictions in fractional shortening (FS) and ejection fraction (EF) evoked by doxorubicin were not amended by the statin. Taken together, the data suggest beneficial effects of lovastatin against doxorubicin-induced delayed cardiotoxicity. Clinical studies are preferable to scrutinize the usefulness of statins for the prevention of anthracycline-induced late cardiotoxicity.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
DL-Serine, ≥98% (TLC)
Sigma-Aldrich
DL-Serine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)
Sigma-Aldrich
Ethanol, absolute, reag. ISO, reag. Ph. Eur., ≥99.8% (GC), liquid (clear, colorless)
Sigma-Aldrich
Ethanol, puriss., meets analytical specification of Ph. Eur., BP, 96.0-97.2%
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.