Merck
  • Preclinical Evaluation of a Potential GSH Ester Based PET/SPECT Imaging Probe DT(GSHMe)₂ to Detect Gamma Glutamyl Transferase Over Expressing Tumors.
Preclinical Evaluation of a Potential GSH Ester Based PET/SPECT Imaging Probe DT(GSHMe)₂ to Detect Gamma Glutamyl Transferase Over Expressing Tumors.
PloS one (2015-07-30)
Harleen Khurana, Virendra Kumar Meena, Surbhi Prakash, Krishna Chuttani, Nidhi Chadha, Ambika Jaswal, Devinder Kumar Dhawan, Anil Kumar Mishra, Puja Panwar Hazari
ABSTRACT
Gamma Glutamyl Transferase (GGT) is an important biomarker in malignant cancers. The redox processes ensuing from GGT-mediated metabolism of extracellular GSH are implicated in critical aspects of tumor cell biology. Reportedly, Glutathione monoethyl ester (GSHMe) is a substrate of GGT, which has been used for its rapid transport over glutathione. Exploring GGT to be an important target, a homobivalent peptide system, DT(GSHMe)2 was designed to target GGT-over expressing tumors for diagnostic purposes. DT(GSHMe)2 was synthesized, characterized and preclinically evaluated in vitro using toxicity, cell binding assays and time dependent experiments. Stable and defined radiochemistry with 99mTc and 68Ga was optimized for high radiochemical yield. In vivo biodistribution studies were conducted for different time points along with scintigraphic studies of radiolabeled DT(GSHMe)2 on xenografted tumor models. For further validation, in silico docking studies were performed on GGT (hGGT1, P19440). Preclinical in vitro evaluations on cell lines suggested minimal toxicity of DT(GSHMe)2 at 100 μM concentration. Kinetic analysis revealed transport of 99mTc-DT(GSHMe)2 occurs via a saturable high-affinity carrier with Michaelis constant (Km) of 2.25 μM and maximal transport rate velocity (Vmax) of 0.478 μM/min. Quantitative estimation of GGT expression from western blot experiments showed substantial expression with 41.6 ± 7.07 % IDV for tumor. Small animal micro PET (Positron Emission Tomography)/CT(Computed Tomography) coregistered images depicted significantly high uptake of DT(GSHMe)2 at the BMG-1 tumor site. ROI analysis showed high tumor to contra lateral muscle ratio of 9.33 in PET imaging studies. Avid accumulation of radiotracer was observed at tumor versus inflammation site at 2 h post i.v. injection in an Ehrlich Ascites tumor (EAT) mice model, showing evident specificity for tumor. We propose DT(GSHMe)2 to be an excellent candidate for prognostication and tumor imaging using PET/SPECT.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)