Merck
  • Hepatocyte growth factor activator is a potential target proteinase for Kazal-type inhibitor in turkey (Meleagris gallopavo) seminal plasma.
Hepatocyte growth factor activator is a potential target proteinase for Kazal-type inhibitor in turkey (Meleagris gallopavo) seminal plasma.
Theriogenology (2015-05-02)
Mariola Słowińska, Joanna Bukowska, Anna Hejmej, Barbara Bilińska, Krzysztof Kozłowski, Jan Jankowski, Andrzej Ciereszko
ABSTRACT
A peculiar characteristic of turkey seminal plasma is the increased activity of serine proteinases. It is of interest if the single-domain Kazal-type inhibitor controls the activity of turkey seminal plasma proteinases. Pure preparations of the Kazal-type inhibitor and anti-Kazal-type inhibitor monospecific immunoglobulin Gs were used as ligands in affinity chromatography for proteinase isolation from turkey seminal plasma. Gene expression and the immunohistochemical detection of the single-domain Kazal-type inhibitor in the reproductive tract of turkey toms are described. The hepatocyte growth factor activator (HGFA) was identified in the binding fraction in affinity chromatography. Hepatocyte growth factor activator activity was inhibited by the Kazal-type inhibitor in a dose-dependent manner. This protease was a primary physiological target for the single-domain Kazal-type inhibitor. Numerous proteoforms of HGFA were present in turkey seminal plasma, and phosphorylation was the primary posttranslational modification of HGFA. In addition to HGFA, acrosin was a target proteinase for the single-domain Kazal-type inhibitor. In seminal plasma, acrosin was present only in complexes with the Kazal-type inhibitor and was not present as a free enzyme. The single-domain Kazal-type inhibitor was specific for the reproductive tract. The germ cell-specific expression of Kazal-type inhibitors in the testis indicated an important function in spermatogenesis; secretion by the epithelial cells of the epididymis and the ductus deferens indicated that the Kazal-type inhibitor was an important factor involved in the changes in sperm membranes during maturation and in the maintenance of the microenvironment in which sperm maturation occurred and sperm was stored. The role of HGFA in these processes remains to be established.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Potassium hydroxide on alumina
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
o-Xylene, anhydrous, 97%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis
Sigma-Aldrich
Potassium hydroxide, AR, pellets, ≥85%
Sigma-Aldrich
Sodium bicarbonate, Hybri-Max, powder, suitable for hybridoma, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Potassium perchlorate, ≥99.99% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, Vetec, reagent grade, ≥98%