Merck
  • A cheap and non-destructive approach to increase coverage/loading of hydrophilic hydroxide on hydrophobic carbon for lightweight and high-performance supercapacitors.
A cheap and non-destructive approach to increase coverage/loading of hydrophilic hydroxide on hydrophobic carbon for lightweight and high-performance supercapacitors.
Scientific reports (2015-12-09)
Liuyang Zhang, Hao Gong
ABSTRACT
Carbon-based substrates offer unprecedented advantages in lightweight supercapacitors. However, it is still challenging to achieve high coverage or loading. Different from the traditional belief that a lack of defects or functional groups is the cause of poor growth on carbon-based substrates, we reckon that the major cause is the discrepancy between the hydrophilic nature of the metal oxide/hydroxide and the hydrophobic nature of carbon. To solve this incompatibility, we introduced ethanol into the precursor solution. The method to synthesize nickel copper hydroxide on carbon fiber paper employs only water and ethanol, in addition to nickel acetate and copper acetate. The results revealed good growth and tight adhesion of active materials on carbon fiber paper substrates. The specific capacitance and energy density per total weight of the active material plus substrate (carbon fiber paper, current collector) reached 770 F g(-1) and 33 Wh kg(-1) (1798 F g(-1) and 54 Wh kg(-1) per weight of the active materials), owing to the high loading of active material and the light weight of carbon fiber paper. These results signified the achievability of light, cheap and high-performance supercapacitors by an environmental-friendly approach.
Sigma-Aldrich
Potassium hydroxide on alumina
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis
Sigma-Aldrich
Potassium hydroxide, puriss. p.a., ≥86% (T), pellets
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, 85-100.5%, pellets
Sigma-Aldrich
Potassium hydroxide, pellets, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
Potassium hydroxide, AR, pellets, ≥85%