Merck
  • MarVis-Pathway: integrative and exploratory pathway analysis of non-targeted metabolomics data.
MarVis-Pathway: integrative and exploratory pathway analysis of non-targeted metabolomics data.
Metabolomics : Official journal of the Metabolomic Society (2015-05-15)
Alexander Kaever, Manuel Landesfeind, Kirstin Feussner, Alina Mosblech, Ingo Heilmann, Burkhard Morgenstern, Ivo Feussner, Peter Meinicke
ABSTRACT
A central aim in the evaluation of non-targeted metabolomics data is the detection of intensity patterns that differ between experimental conditions as well as the identification of the underlying metabolites and their association with metabolic pathways. In this context, the identification of metabolites based on non-targeted mass spectrometry data is a major bottleneck. In many applications, this identification needs to be guided by expert knowledge and interactive tools for exploratory data analysis can significantly support this process. Additionally, the integration of data from other omics platforms, such as DNA microarray-based transcriptomics, can provide valuable hints and thereby facilitate the identification of metabolites via the reconstruction of related metabolic pathways. We here introduce the MarVis-Pathway tool, which allows the user to identify metabolites by annotation of pathways from cross-omics data. The analysis is supported by an extensive framework for pathway enrichment and meta-analysis. The tool allows the mapping of data set features by ID, name, and accurate mass, and can incorporate information from adduct and isotope correction of mass spectrometry data. MarVis-Pathway was integrated in the MarVis-Suite (http://marvis.gobics.de), which features the seamless highly interactive filtering, combination, clustering, and visualization of omics data sets. The functionality of the new software tool is illustrated using combined mass spectrometry and DNA microarray data. This application confirms jasmonate biosynthesis as important metabolic pathway that is upregulated during the wound response of Arabidopsis plants.
Sigma-Aldrich
L-Isoleucine, 99%, FCC, FG
Sigma-Aldrich
L-Isoleucine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Isoleucine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)