Merck
  • Droplet spray ionization from a glass microscope slide: real-time monitoring of ethylene polymerization.
Droplet spray ionization from a glass microscope slide: real-time monitoring of ethylene polymerization.
Analytical chemistry (2015-07-24)
Jie Jiang, Hong Zhang, Ming Li, Maria T Dulay, Andrew J Ingram, Na Li, Hong You, Richard N Zare
ABSTRACT
Ambient ionization mass spectrometry is achieved in a simple manner by loading a sample solution onto a corner of a microscope cover glass positioned in front of the inlet to a mass spectrometer and applying a high voltage to the sample. The resulting stream of charged droplets is stable, has no contamination from the substrate platform, and can be used repeatedly. The utility of droplet spray for in situ analysis and real-time monitoring of chemical reactions was demonstrated by the bis(cyclopentadienyl)zirconium dichloride (zirconocene dichloride)/methylaluminoxane, Cp2ZrCl2/MAO, homogeneously catalyzed polymerization of ethylene in various solutions. Reaction times ranged from seconds to minutes, and catalytically active species and polymeric products of ethylene were acquired and identified by tandem mass spectrometry.
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Tetrahydrofuran, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Fluorobenzene, 99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)