Merck
  • Increase in acrolein-conjugated immunoglobulins in saliva from patients with primary Sjögren's syndrome.
Increase in acrolein-conjugated immunoglobulins in saliva from patients with primary Sjögren's syndrome.
Clinica chimica acta; international journal of clinical chemistry (2015-08-25)
Tadao Hirose, Ryotaro Saiki, Takeshi Uemura, Takehiro Suzuki, Naoshi Dohmae, Satoshi Ito, Hoyu Takahashi, Itsuko Ishii, Toshihiko Toida, Keiko Kashiwagi, Kazuei Igarashi
ABSTRACT
We previously reported that the level of protein-conjugated acrolein (PC-Acro), a marker of cell or tissue damage, was increased in saliva from patients with primary Sjögren's syndrome (pSS), and that the level of PC-Acro was well correlated with the severity of pSS. Acrolein-conjugated immunoglobulins were measured in saliva from pSS patients. The activities of autoantibodies recognizing Sjögren's syndrome SSA (Ro) and SSB (La) proteins in saliva from pSS patients were approximately 3- to 5-fold higher than those from control subjects. We also found that autoantibody activities recognizing SSA (Ro) and SSB (La) proteins increased after acrolein treatment of saliva from control subjects. When an antibody against human serum albumin was treated with acrolein, the ability to recognize albumin was reduced but the ability to recognize other proteins was increased. Twenty-four and eleven kinds of acrolein-conjugated amino acids were found at the variable and constant regions of peptides, respectively, obtained from the immunoglobulins in saliva from pSS patients. The altered recognition patterns of immunoglobulins due to acrolein conjugation are at least partially involved in autoimmune diseases.
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Acrolein, analytical standard
Sigma-Aldrich
Acrolein, contains hydroquinone as stabilizer, 90%