Merck
  • C-terminal incorporation of fluorogenic and affinity labels using wild-type and mutagenized carboxypeptidase Y.
C-terminal incorporation of fluorogenic and affinity labels using wild-type and mutagenized carboxypeptidase Y.
Analytical biochemistry (1997-05-15)
H R Stennicke, K Olesen, S B Sørensen, K Breddam
ABSTRACT
The ability to carry out specific C-terminal modification or labeling of peptides and proteins has a broad range of applications. It is well established that this may be achieved by protease-catalyzed transacylation reactions and that carboxypeptidase Y (CPD-Y) is suitable for this due to its broad specificity and stability in the presence of denaturants. Furthermore, CPD-Y is characterized by a S'1 binding site that is open to solvent and, thus, capable of catalyzing a transpeptidation reaction with nucleophiles that extend beyond the perimeter of the active site. However, one major drawback with CPD-Y is that the yield of the reaction is highly dependent on the nature of the leaving group; e.g., with large apolar leaving groups the yield of the reaction does not exceed 15%. In the present publication it is demonstrated that mutants of CPD-Y, designed for low leaving group dependence, efficiently incorporate biocytin amide as well as a new fluorescent nucleophile, N'-Abz-Lysine amide (ablysin amide), into peptides and proteins.
Sigma-Aldrich
Biocytinamide