Merck
  • The novel CGRP receptor antagonist BIBN4096BS alleviates a postoperative intestinal inflammation and prevents postoperative ileus.
The novel CGRP receptor antagonist BIBN4096BS alleviates a postoperative intestinal inflammation and prevents postoperative ileus.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society (2015-05-02)
T R Glowka, A Steinebach, K Stein, T Schwandt, M Lysson, B Holzmann, K Tsujikawa, W J de Jonge, J C Kalff, S Wehner
ABSTRACT
Abdominal surgery results in neuronal mediator release and subsequent acute intestinal hypomotility. This phase is followed by a longer lasting inflammatory phase resulting in postoperative ileus (POI). Calcitonin gene-related peptide (CGRP) has been shown to induce motility disturbances and in addition may be a candidate mediator to elicit neurogenic inflammation. We hypothesized that CGRP contributes to intestinal inflammation and POI. The effect of CGRP in POI was tested in mice treated with the highly specific CGRP receptor antagonist BIBN4096BS and in CGRP receptor-deficient (RAMP-1(-/-) ) mice. POI severity was analyzed by cytokine expression, muscular inflammation and gastrointestinal (GI) transit. Peritoneal and muscularis macrophages and mast cells were analyzed for CGRP receptor expression and functional response to CGRP stimulation. Intestinal manipulation (IM) resulted in CGRP release from myenteric nerves, and a concurrent increased interleukin (IL)-6 and IL-1β transcription and leukocyte infiltration in the muscularis externa and increased GI transit time. CGRP potentiates IM-induced cytokine transcription within the muscularis externa and peritoneal macrophages. BIBN4096BS reduced cytokine levels and leukocyte infiltration and normalized GI transit. RAMP1(-/-) mice showed a significantly reduced leukocyte influx. CGRP receptor was expressed in muscularis and peritoneal macrophages but not mast cells. CGRP mediated macrophage activation but failed to induce mast cell degranulation and cytokine expression. CGRP is immediately released during abdominal surgery and induces a neurogenic inflammation via activation of abdominal macrophages. BIBN4096BS prevented IM-induced inflammation and restored GI motility. These findings suggest that CGRP receptor antagonism could be instrumental in the prevention of POI.
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Capsaicin, ≥95%, from Capsicum sp.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
1,2,4,5-Tetramethylbenzene, 95%
Sigma-Aldrich
Capsaicin, natural
Sigma-Aldrich
Capsaicin, from Capsicum sp., ≥50% (HPLC)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
L-Glutamine, Vetec, reagent grade, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, ACS reagent, prima fine spirit, without additive, F15 o1
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 1% cyclohexane, A15 CYCLO1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, puriss. p.a., absolute, ≥99.8% (GC)