Merck
  • High-dose Ionizing Radiation Regulates Gene Expression Changes in the MCF7 Breast Cancer Cell Line.
High-dose Ionizing Radiation Regulates Gene Expression Changes in the MCF7 Breast Cancer Cell Line.
Anticancer research (2015-05-13)
Valentina Bravatà, Luigi Minafra, Giorgio Russo, Giusi Irma Forte, Francesco P Cammarata, Marilena Ripamonti, Carlo Casarino, Giuseppa Augello, Francesca Costantini, Giovanna Barbieri, Cristina Messa, Maria C Gilardi
ABSTRACT
Intraoperative electron radiation therapy (IOERT) is a therapeutic technique which administers a single high dose of ionizing radiation immediately after surgical tumor removal. IOERT induces a strong stress response: both tumor and normal cells activating pro- and antiproliferative cell signaling pathways. Following treatment, several genes and factors are differently modulated, producing an imbalance in cell fate decision. However, the contribution of these genes and pathways in conferring different cell radiosensitivity and radioresistance needs to be further investigated, in particular after high-dose treatments. Despite the documented and great impact of IOERT in breast cancer care, and the trend for dose escalation, very limited data are available regarding gene-expression profiles and cell networks activated by IOERT or high-dose treatment. The aim of the study was to analyze the main pathways activated following high radiation doses in order to select for potential new biomarkers of radiosensitivity or radioresistance, as well as to identify therapeutic targets useful in cancer care. We performed gene-expression profiling of the MCF7 human breast carcinoma cell line after treatment with 9- and 23-Gy doses (conventionally used during IOERT boost and exclusive treatments, respectively) by cDNA microarrays. Real-Time Quantitative Reverse Transcription PCR (qRT-PCR), immunofluorescence and immunoblot experiments were performed to validate candidate IOERT biomarkers. We also conducted clonogenic tests and cellular senescence assays to monitor for radiation-induced effects. The analyses highlighted a transcriptome dependent on the dose delivered and a number of specific key genes that may be proposed as new markers of radiosensitivity. Cell and molecular traits observed in MCF7 cells revealed a typical senescent phenotype associated with cell proliferation arrest after treatments with 9- and 23-Gy doses. In this study, we report genes and cellular networks activated following high-dose IOERT. The selected validated genes were used to design two descriptive models for each dose delivered. We believe that this study could contribute to the understanding over the complex mechanisms which regulate cell radiosensitivity and radioresistance in order to improve personalized radiotherapeutic treatment.
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
3-Maleimidobenzoic acid N-hydroxysuccinimide ester, crystalline
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Adenosine 5′-diphosphoribose sodium salt, ≥93%
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture