Merck
  • N-Myristoylation of the Rpt2 subunit of the yeast 26S proteasome is implicated in the subcellular compartment-specific protein quality control system.
N-Myristoylation of the Rpt2 subunit of the yeast 26S proteasome is implicated in the subcellular compartment-specific protein quality control system.
Journal of proteomics (2015-09-08)
Ayuko Kimura, Yoichi Kurata, Jun Nakabayashi, Hiroyuki Kagawa, Hisashi Hirano
ABSTRACT
Ubiquitination is the posttranslational modification of a protein by covalent attachment of ubiquitin. Controlled proteolysis via the ubiquitin-proteasome system (\UPS) alleviates cellular stress by clearing misfolded proteins. In budding yeast, UPS within the nucleus degrades the nuclear proteins as well as proteins imported from the cytoplasm. While the predominantly nuclear localization of the yeast proteasome is maintained by the importin-mediated transport, N-myristoylation of the proteasome subunit Rpt2 was indicated to cause dynamic nucleo-cytoplasmic localization of proteasomes. Here, we quantitatively analyzed the ubiquitinated peptides using anti-K-ε-GG antibody in yeast cell lines with or without a mutation in the N-myristoylation site of Rpt2 and detected upregulated ubiquitination of proteins with nucleo-cytoplasmic localizations in the mutant strains. Moreover, both the protein and ubiquitinated peptide levels of two Hsp70 family chaperones involved in the nuclear import of misfolded proteins, Ssa and Sse1, were elevated in the mutant strains, whereas levels of an Hsp70 family chaperone involved in the nuclear export, Ssb, were reduced. Taken together, our results indicate that N-myristoylation of Rpt2 is involved in controlled proteolysis via regulation of the nucleo-cytoplasmic localization of the yeast proteasome.
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
2-Chloroacetamide, ≥98.0% (HPLC)
Sigma-Aldrich
2-Chloroacetamide, ≥98%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
HEPES, Vetec, reagent grade, 99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Urea, Vetec, reagent grade, 99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, Vetec, reagent grade, 98%
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder