Merck
  • Separation of the intracellular secretory compartment of rat liver and isolated rat hepatocytes in a single step using self-generating gradients of iodixanol.
Separation of the intracellular secretory compartment of rat liver and isolated rat hepatocytes in a single step using self-generating gradients of iodixanol.
Analytical biochemistry (1999-12-10)
D Plonne, I Cartwright, W Linss, R Dargel, J M Graham, J A Higgins
ABSTRACT
A novel method is described for the separation on a single gradient of the major intracellular organelles of the secretory pathway, the Golgi, the smooth endoplasmic reticulum, and the rough endoplasmic reticulum. Total microsomes were prepared from rat liver by differential centrifugation and resuspended in 20% iodixanol. The microsomal suspension was then layered between a 30% iodixanol cushion and a layer of 15% iodixanol and centrifuged in a vertical rotor for 2 h. The microsomes distributed in four visible bands. The gradients were collected by upward displacement and were characterized (i) by determination of UDP galactose-galactosyltransferase (Golgi marker) NADPH-cytochrome c reductase (endoplasmic reticulum marker) and RNA (rough endoplasmic reticulum marker); (ii) by immunoblotting for TGN38 (trans-Golgi marker) and GS28 (cis-Golgi marker) and for protein disulfide isomerase (endoplasmic reticulum lumenal marker); (iii) by determination of the lipid composition; and (iv) by electron microscopy. The results suggest that the top band (density 1.045-1. 090 g/ml), which contains 68% of the galactosyltransferase activity, consists of vesicles derived from the Golgi. The second broad band in the middle of the tube (density 1.130-1.160 g/ml), which contains 54% of the NADPH-cytochrome c reductase activity, consists mainly of vesicles derived from the smooth endoplasmic reticulum, overlapped at the top by a small band of Golgi-derived lamellae. The two bands at the bottom of the tube (density 1.130-1.160 and density 1.180-1. 220 g/ml) appear to contain two subfractions of vesicles derived from the rough endoplasmic reticulum.
Sigma-Aldrich
Cytochrome c Reductase (NADPH) Assay Kit, 1 kit sufficient for 100 tests, determining cytochrome c reductase activity
Pricing and availability is not currently available.