Merck
  • Aging without Apolipoprotein D: Molecular and cellular modifications in the hippocampus and cortex.
Aging without Apolipoprotein D: Molecular and cellular modifications in the hippocampus and cortex.
Experimental gerontology (2015-04-15)
Diego Sanchez, Raquel Bajo-Grañeras, Manuela Del Caño-Espinel, Rosa Garcia-Centeno, Nadia Garcia-Mateo, Raquel Pascua-Maestro, Maria D Ganfornina
ABSTRACT
A detailed knowledge of the mechanisms underlying brain aging is fundamental to understand its functional decline and the baseline upon which brain pathologies superimpose. Endogenous protective mechanisms must contribute to the adaptability and plasticity still present in the healthy aged brain. Apolipoprotein D (ApoD) is one of the few genes with a consistent and evolutionarily conserved up-regulation in the aged brain. ApoD protecting roles upon stress or injury are well known, but a study of the effects of ApoD expression in the normal aging process is still missing. Using an ApoD-knockout mouse we analyze the effects of ApoD on factors contributing to the functional maintenance of the aged brain. We focused our cellular and molecular analyses in the cortex and hippocampus at an age representing the onset of senescence where mortality risks are below 25%, avoiding bias towards long-lived animals. Lack of ApoD causes a prematurely aged brain without altering lifespan. Age-dependent hyperkinesia and memory deficits are accompanied by differential molecular effects in the cortex and hippocampus. Transcriptome analyses reveal distinct effects of ApoD loss on the molecular age-dependent patterns of the cortex and hippocampus, with different cell-type contributions to age-regulated gene expression. Markers of glial reactivity, proteostasis, and oxidative and inflammatory damage reveal early signs of aging and enhanced brain deterioration in the ApoD-knockout brain. The lack of ApoD results in an age-enhanced significant reduction in neuronal calcium-dependent functionality markers and signs of early reduction of neuronal numbers in the cortex, thus impinging upon parameters clearly differentiating neurodegenerative conditions from healthy brain aging. Our data support the hypothesis that the physiological increased brain expression of ApoD represents a homeostatic anti-aging mechanism.
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
Sodium, 99.95% trace metals basis, ingot
Sigma-Aldrich
Potassium, cubes (in mineral oil), 99.5% trace metals basis
Sigma-Aldrich
Potassium hydride, in paraffin
Sigma-Aldrich
Guanosine 5′-diphospho-D-mannose sodium salt from Saccharomyces cerevisiae, Type I, ≥97% (HPLC)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
DL-Leucine, ≥99% (HPLC)
Sigma-Aldrich
Spermine, ≥97%
Sigma-Aldrich
DL-Arginine, ≥95% (NT)
Sigma-Aldrich
Potassium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Potassium hydride, 30 wt % dispersion in mineral oil
Sigma-Aldrich
Spermine, ≥99.0% (GC)
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Arsenic, 99.999% trace metals basis
Sigma-Aldrich
Sodium, 25-35 wt % dispersion in paraffin
Sigma-Aldrich
Spermine, suitable for cell culture, BioReagent
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
Arsenic, powder, ≥99.997% trace metals basis
Sigma-Aldrich
Potassium, ingot, 99.95% trace metals basis
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.5% trace metals basis
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
Spermidine, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Spermidine, BioReagent, Molecular Biology, suitable for cell culture, ≥98%
Sigma-Aldrich
Sodium, in kerosene, pieces (large), ≥99.8% (sodium basis)
Sigma-Aldrich
Sodium, ACS reagent, dry