Merck
  • Target-induced reconfiguration of DNA probes for recycling amplification and signal-on electrochemical detection of hereditary tyrosinemia type I gene.
Target-induced reconfiguration of DNA probes for recycling amplification and signal-on electrochemical detection of hereditary tyrosinemia type I gene.
The Analyst (2015-07-17)
Baoting Dou, Cuiyun Yang, Yaqin Chai, Ruo Yuan, Yun Xiang
ABSTRACT
By coupling target DNA-induced reconfiguration of the dsDNA probes with enzyme-assisted target recycling amplification, we describe the development of a signal-on electrochemical sensing approach for sensitive detection of hereditary tyrosinemia type I gene. The dsDNA probes are self-assembled on the sensing electrode, and the addition of the target DNA reconfigures and switches the dsDNA probes into active substrates for exonuclease III, which catalytically digests the probes and leads to cyclic reuse of the target DNA. The target DNA recycling and the removal of one of the ssDNA from the dsDNA probes by exonuclease III result in the formation of many hairpin structures on the sensor surface, which brings the electroactive methylene blue labels into proximity with the electrode and produces a significantly amplified current response for sensitive detection of the target gene down to 0.24 pM. This method is also selective to discriminate single-base mismatch and can be employed to detect the target gene in human serum samples. With the demonstration for the detection of the target gene, we expect the developed method to be a universal sensitive sensing platform for the detection of different nucleic acid sequences.
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Supelco
Tris(2-carboxyethyl)phosphine hydrochloride solution, 0.5 M, pH 7.0(aqueous solution; pH was adjusted with ammonium hydroxide)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, suitable for electrophoresis, SDS-PAGE tested
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
6-Mercapto-1-hexanol, 99%
Sigma-Aldrich
6-Mercapto-1-hexanol, 97%
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
1-Hexanethiol, 95%
Sigma-Aldrich
1-Hexanethiol, 99% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O