Merck
  • Comparative proteomics of Shigella flexneri 2a strain 301 using a rabbit ileal loop model reveals key proteins for bacterial adaptation in host niches.
Comparative proteomics of Shigella flexneri 2a strain 301 using a rabbit ileal loop model reveals key proteins for bacterial adaptation in host niches.
International journal of infectious diseases : IJID : official publication of the International Society for Infectious Diseases (2015-09-30)
Xingming Liu, Lilan Lu, Xiankai Liu, Chao Pan, Erling Feng, Dongshu Wang, Li Zhu, Hengliang Wang
ABSTRACT
Many studies focusing on changes in the host following Shigella spp invasion have been reported in recent years. However, the key factors required for the adaptation of these pathogens to host niches have usually been neglected. In this study, a comparative proteomic analysis was performed to examine changes in the protein expression profile of Shigella flexneri within the host using a rabbit ileal loop model to reveal proteins that are associated with pathogenic adaptation. The protein expression profiles of bacteria isolated from the ileum and colon were very similar, although they differed slightly from that of bacteria isolated from the cecum. When compared with the sample in vitro, the expressions of seven proteins were found to be upshifted in vivo (OmpA, YgiW, MglB, YfiD, MetK, TktA, and AhpF), while two proteins were down-regulated (ElaB and GlnH). The abundance of nine proteins changed in vivo, suggesting that these proteins may contribute to adaptation to the intestinal lumen.
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 97%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, suitable for MALDI-TOF MS
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, 99%
Sigma-Aldrich
α-Cyano-4-hydroxycinnamic acid, ≥98% (TLC), powder
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC