Merck
  • Inhibition of endothelial cell migration by thrombospondin-1 type-1 repeats is mediated by beta1 integrins.
Inhibition of endothelial cell migration by thrombospondin-1 type-1 repeats is mediated by beta1 integrins.
The Journal of cell biology (2005-02-18)
Sarah M Short, Alexandrine Derrien, Radha P Narsimhan, Jack Lawler, Donald E Ingber, Bruce R Zetter
ABSTRACT
The anti-angiogenic effect of thrombospondin-1 has been shown to be mediated through binding of the type-1 repeat (TSR) domain to the CD36 transmembrane receptor. We now report that the TSR domain can inhibit VEGF-induced migration in human umbilical vein endothelial cells (HUVEC), cells that lack CD36. Moreover, we identified beta1 integrins as a critical receptor in TSR-mediated inhibition of migration in HUVEC. Using pharmacological inhibitors of downstream VEGF receptor effectors, we found that phosphoinositide 3-kinase (PI3k) was essential for TSR-mediated inhibition of HUVEC migration, but that neither PLCgamma nor Akt was necessary for this response. Furthermore, beta1 integrins were critical for TSR-mediated inhibition of microvascular endothelial cells, cells that express CD36. Together, our results indicate that beta1 integrins mediate the anti-migratory effects of TSR through a PI3k-dependent mechanism.
Sigma-Aldrich
Anti-Integrin β1 antibody, Mouse monoclonal, clone W1B10, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone HB1.1, clone HB1.1, Chemicon®, from mouse
Pricing and availability is not currently available.