Merck
  • Synthesis and structure-activity relationship of aminobenzophenones. A novel class of p38 MAP kinase inhibitors with high antiinflammatory activity.
Synthesis and structure-activity relationship of aminobenzophenones. A novel class of p38 MAP kinase inhibitors with high antiinflammatory activity.
Journal of medicinal chemistry (2003-12-12)
Erik Rytter Ottosen, Morten Dahl Sørensen, Fredrik Björkling, Tine Skak-Nielsen, Marianne Scheel Fjording, Helle Aaes, Lise Binderup
ABSTRACT
We wish to report the synthesis and structure-activity relationship (SAR) of a series of 4-aminobenzophenones, as a novel compound class with high antiinflammatory activity. Our initial lead, (4-[(2-aminophenyl)amino]phenyl)(phenyl)methanone (3), was systematically optimized and resulted in compounds that potently inhibited the release of the proinflammatory cytokines IL-1beta and TNF-alpha in human peripheral blood mononuclear cells stimulated by LPS. One of the most potent compounds, among others, was (4-[(2-aminophenyl)amino]-2-chlorophenyl)(2-methylphenyl)methanone (45) with IC(50) values of 14 and 6 nM for the inhibition of IL-1beta and TNF-alpha, respectively. Furthermore, we found these types of compounds to be potent and selective p38 MAP kinase inhibitors, e.g. 45 had an IC(50) value of 10 nM. Molecular modeling was used to rationalize our SAR data and to propose a model for the interaction of compound 45 with the p38 MAP kinase. The model involved a favorable hydrogen bond between the carbonyl group of the benzophenone and the NH of Met-109, positioning ring A in the hydrophobic pocket I of the enzyme. Good antiinflammatory effects were demonstrated in two murine models of dermatitis after topical application (oxazolone and TPA model).
Sigma-Aldrich
4-Bromo-1-fluoro-2-nitrobenzene, 96%
Pricing and availability is not currently available.