Merck
Volatiles of Chrysanthemum zawadskii var. latilobum K.
Preventive nutrition and food science (2012-09-01)
Kyung-Mi Chang, Gun-Hee Kim
ABSTRACT
The volatile aroma constituents of Chrysanthemum zawadskii var. latilobum K. were separated by hydro distillation extraction (HDE) method using a Clevenger-type apparatus, and analyzed by gas chromatography-mass spectrometry (GC/MS). The yield of C. zawadskii var. latilobum K. flower essential oil (FEO) was 0.12% (w/w) and the color was light green. Fifty-five volatile chemical components, which make up 88.38% of the total aroma composition, were tentatively characterized. C. zawadskii var. latilobum K. FEOs contained 27 hydrocarbons, 12 alcohols, 7 ketones, 4 esters, 1 aldehyde, 1 amine, and 3 miscellaneous components. The major functional groups were terpene alcohol and ketone. Borneol (12.96), (±)-7-epi-amiteol (12.60), and camphor (10.54%) were the predominant volatiles. These compounds can be used in food and pharmaceutical industries due to their active bio-functional properties.
Product Description
Supelco
Alkane standard solution C8-C20, analytical standard, contains C8-C20, ~40 mg/L each, in hexane
Pricing and availability is not currently available.