Merck
Gene expression-based screening for inhibitors of PDGFR signaling.
Genome biology (2008-03-04)
Alena A Antipova, Brent R Stockwell, Todd R Golub
ABSTRACT
Here we describe a proof-of-concept experiment designed to explore the possibility of using gene expression-based high-throughput screening (GE-HTS) to find inhibitors of a signaling cascade, using platelet derived growth factor receptor (PDGFR) signaling as the example. The previously unrecognized ability of aurintricarboxylic acid to inhibit PDGFR signaling, discovered through a screen of 1,739 compounds, demonstrates the feasibility and generalizability of GE-HTS for the discovery of small molecule modulators of any signaling pathway of interest.
Sigma-Aldrich
Rhodamine 19 perchlorate, Dye content ~95 %
Sigma-Aldrich
3-Methylsalicylic acid, 96.0%