Merck
  • A new class of plant lipid is essential for protection against phosphorus depletion.
A new class of plant lipid is essential for protection against phosphorus depletion.
Nature communications (2013-02-28)
Yozo Okazaki, Hitomi Otsuki, Tomoko Narisawa, Makoto Kobayashi, Satoru Sawai, Yukiko Kamide, Miyako Kusano, Toshio Aoki, Masami Yokota Hirai, Kazuki Saito
ABSTRACT
Phosphorus supply is a major factor responsible for reduced crop yields. As a result, plants utilize various adaptive mechanisms against phosphorus depletion, including lipid remodelling. Here we report the involvement of a novel plant lipid, glucuronosyldiacylglycerol, against phosphorus depletion. Lipidomic analysis of Arabidopsis plants cultured in phosphorus-depleted conditions revealed inducible accumulation of glucuronosyldiacylglycerol. Investigation using a series of sulfolipid sulfoquinovosyldiacylglycerol synthesis-deficient mutants of Arabidopsis determined that the biosynthesis of glucuronosyldiacylglycerol shares the pathway of sulfoquinovosyldiacylglycerol synthesis in chloroplasts. Under phosphorus-depleted conditions, the Arabidopsis sqd2 mutant, which does not accumulate either sulfoquinovosyldiacylglycerol or glucuronosyldiacylglycerol, was the most severely damaged of three sulfoquinovosyldiacylglycerol-deficient mutants. As glucuronosyldiacylglycerol is still present in the other two mutants, this result indicates that glucuronosyldiacylglycerol has a role in the protection of plants against phosphorus limitation stress. Glucuronosyldiacylglycerol was also found in rice, and its concentration increased significantly following phosphorus limitation, suggesting a shared physiological significance of this novel lipid against phosphorus depletion in plants.
Sigma-Aldrich
1,2-Didecanoyl-sn-glycero-3-phosphocholine, ≥99% (TLC)