Merck
  • Single-molecule spectroscopy of protein conformational dynamics in live eukaryotic cells.
Single-molecule spectroscopy of protein conformational dynamics in live eukaryotic cells.
Nature methods (2015-07-07)
Iwo König, Arash Zarrine-Afsar, Mikayel Aznauryan, Andrea Soranno, Bengt Wunderlich, Fabian Dingfelder, Jakob C Stüber, Andreas Plückthun, Daniel Nettels, Benjamin Schuler
ABSTRACT
Single-molecule methods have become widely used for quantifying the conformational heterogeneity and structural dynamics of biomolecules in vitro. Their application in vivo, however, has remained challenging owing to shortcomings in the design and reproducible delivery of labeled molecules, the range of applicable analysis methods, and suboptimal cell culture conditions. By addressing these limitations in an integrated approach, we demonstrate the feasibility of probing protein dynamics from milliseconds down to the nanosecond regime in live eukaryotic cells with confocal single-molecule Förster resonance energy transfer (FRET) spectroscopy. We illustrate the versatility of the approach by determining the dimensions and submicrosecond chain dynamics of an intrinsically disordered protein; by detecting even subtle changes in the temperature dependence of protein stability, including in-cell cold denaturation; and by quantifying the folding dynamics of a small protein. The methodology opens possibilities for assessing the effect of the cellular environment on biomolecular conformation, dynamics and function.
Sigma-Aldrich
Abberior® STAR 635, maleimide, for STED application