Merck
  • Diastereocontrolled synthesis of enantioenriched 3,4-disubstituted beta-prolines.
Diastereocontrolled synthesis of enantioenriched 3,4-disubstituted beta-prolines.
The Journal of organic chemistry (2007-01-16)
Fabrice Denes, Alejandro Perez-Luna, Fabrice Chemla
ABSTRACT
Enantioenriched 3,4-disubstituted beta-prolines have been prepared with a high diastereocontrol through a carbometalation reaction or through a domino Michael addition/carbometalation reaction.
Sigma-Aldrich
(R)-(−)-Pyrrolidine-3-carboxylic acid, ≥99.0% (NT)
Sigma-Aldrich
Methyl 2-(bromomethyl)acrylate, 97%
Sigma-Aldrich
(S)-(+)-Pyrrolidine-3-carboxylic acid, ≥98.0% (NT)