- Cis double allylation of cyclopropenes using cyclopropylindium reagents.
Cis double allylation of cyclopropenes using cyclopropylindium reagents.
Organic & biomolecular chemistry (2007-06-22)
Tsunehisa Hirashita, Fumio Shiraki, Kenji Onishi, Mamiko Ogura, Shuki Araki
PMID17581660
ABSTRACT
The successive double allylation of cyclopropenes with allylindium sesquiiodide and allyl iodide proceeded with a cis-addition mode in the presence of other organometallics (e.g. Grignard reagent, cuprate, Et(2)Zn and Et(3)Al), giving the corresponding cis-diallylcyclopropanes in high yields.